Learn More
Medchemexpress LLC ADP-Glucose (disodium) | 102129-65-7 | 95.0% | 633.31 | 50 MG
SDP
Catalog No. 5000348698
Encompass_Preferred
ADP-Glucose disodium is an immediate precursor for the biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria, and structural polysaccharides in some bacteria. It is utilized to produce amylose, amylopectin, starch, and other polysaccharides by amylose synthase or starch synthase in plastids. This product is for research use only.
  • Precursor for biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria.
  • Precursor for structural polysaccharides in some bacteria.
  • Utilized for the production of amylose, amylopectin, and starch.
  • Produced in plastids and can be imported via membrane-bound transporters.
  • Suitable for laboratory chemical applications.
  • For research use only.
Catalog No. 50-003-48698 Supplier Medchemexpress LLC Supplier No. HY13683050MG
ADP-Glucose disodium is an immediate precursor for the biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria, and structural polysaccharides in some bacteria. It is utilized to produce amylose, amylopectin, starch, and other polysaccharides by amylose synthase or starch synthase in plastids. This product is for research use only.
  • Precursor for biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria.
  • Precursor for structural polysaccharides in some bacteria.
  • Utilized for the production of amylose, amylopectin, and starch.
  • Produced in plastids and can be imported via membrane-bound transporters.
  • Suitable for laboratory chemical applications.
  • For research use only.