Learn More
Cayman Chemical ADP-Glucose (sodium salt), Purity: ≥95%, CAS: 102129-65-7, 10 mg
SDP
Catalog No. 502966683
Encompass_Preferred
An immediate precursor used in the biosynthesis, by glucose addition, of storage polysaccharides in plants, green algae, and cyanobacteria, as well as structural polysaccharides in certain bacteria
Catalog No. 50-296-6683 Supplier Cayman Chemical Supplier No. 1813910 MG
An immediate precursor used in the biosynthesis, by glucose addition, of storage polysaccharides in plants, green algae, and cyanobacteria, as well as structural polysaccharides in certain bacteria