Learn More
Eurofins Discoverx ADP Hunter Plus, 100 (10,000 dp, 384-well). (90-0083L)
Catalog No. 501999535
Encompass
The ADP Hunter Plus kit is a detection kit for the measurement of ADP produced as a product of kinase activity. It is used to detect substrate-induced activity of purified or semi-purified kinases. ADP produced by the kinase reacts with intermediate factors to produce a fluorescent dye precursor, which generates signal when combined with peroxide and peroxidase. The kit uses an end-point read. This detection kit contains reagents for 25 plates (384-well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.
Catalog No. 50-199-9535 Supplier Eurofins Discoverx Supplier No. 900083L
The ADP Hunter Plus kit is a detection kit for the measurement of ADP produced as a product of kinase activity. It is used to detect substrate-induced activity of purified or semi-purified kinases. ADP produced by the kinase reacts with intermediate factors to produce a fluorescent dye precursor, which generates signal when combined with peroxide and peroxidase. The kit uses an end-point read. This detection kit contains reagents for 25 plates (384-well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.