Learn More
Enzo Life Sciences Adenosine 5'-diphosphate. potassium salt (1g). CAS: 72696-48-1
SDP
Catalog No. 502009298
Encompass_Preferred
Alternative name: ADP . K. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9298 Supplier Enzo Life Sciences Supplier No. ALX480002G001
Add to Cart
Add to Cart
Alternative name: ADP . K. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.