Learn More
VAZYME INTERNATIONAL LLC ADP-Lite Kinase Assay 400 Tests
Supplier:  VAZYME INTERNATIONAL LLC DD123001
Encompass
ADP-Lite Kinase Assay is a universal kinase activity assay kit designed for measuring the activity of any ADP-generating enzyme (e.g., kinases and ATPases). To accurately determine target kinase activity, the assay quantifies the ADP produced during a kinase reaction, which is positively correlated with the kinase activity. This simple, rapid, and homogeneous assay is based on a highly sensitive in vitro bioluminescence technology that requires no excitation light or radioactive materials. It is compatible with a wide range of kinase substrates, including lipids, peptides, proteins, and carbohydrates.The kit contains two core components: ADP-Lite Reagent (for ATP depletion) and Kinase Detection Reagent (for ADP detection). Kinase Detection Reagent is prepared by mixing Kinase Detection Buffer and Kinase Detection Substrate (Lyophilized).
Catalog No. NC3979683