Learn More
VAZYME INTERNATIONAL LLC Vazyme International ADP-Lite Kinase Assay, Universal Kinase Activity, 10000 tests
Catalog No. NC3979685
Encompass
The ADP-Lite Kinase Assay is a universal kinase activity assay kit designed for measuring the activity of any ADP-generating enzyme, such as kinases and ATPases.
  • Designed for measuring the activity of any ADP-generating enzyme.
  • Suitable for kinases and ATPases.
  • Kit size: 10000 tests.
Catalog No. NC3979685 Supplier VAZYME INTERNATIONAL LLC Supplier No. DD123003
The ADP-Lite Kinase Assay is a universal kinase activity assay kit designed for measuring the activity of any ADP-generating enzyme, such as kinases and ATPases.
  • Designed for measuring the activity of any ADP-generating enzyme.
  • Suitable for kinases and ATPases.
  • Kit size: 10000 tests.