Learn More
Trajan Scientific ADP OGFS-16-005 10PK
SDP
Catalog No. 502530214
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, For Use With: GC Chromatography Used with Injectors and Detectors at Atmospheric Pressure, Fits Column I.D. 0.01 to 0.032, For Column I.D. 0.1 to 0.32mm, Diameter Inside: 0.019 in., 0.05 cm, Temperature Max: 450 deg.C
Catalog No. 50-253-0214 Supplier Trajan Scientific Supplier No. 072635
Trajan SGE 100% Graphite Ferrules, For Use With: GC Chromatography Used with Injectors and Detectors at Atmospheric Pressure, Fits Column I.D. 0.01 to 0.032, For Column I.D. 0.1 to 0.32mm, Diameter Inside: 0.019 in., 0.05 cm, Temperature Max: 450 deg.C