Learn More
Trajan Scientific ADP OGFS-16-008 10PK
SDP
Catalog No. 502530213
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, For Use With: GC Used with Injectors and Detectors at Atmospheric Pressure, Fits Column I.D. 0.045 to 0.053 cm, For Column I.D. 0.45 to 0.53mm, Diameter Inside: 0.08 cm, Temperature Max: 450 deg.C
Catalog No. 50-253-0213 Supplier Trajan Scientific Supplier No. 072636
Trajan SGE 100% Graphite Ferrules, For Use With: GC Used with Injectors and Detectors at Atmospheric Pressure, Fits Column I.D. 0.045 to 0.053 cm, For Column I.D. 0.45 to 0.53mm, Diameter Inside: 0.08 cm, Temperature Max: 450 deg.C