Learn More
Medchemexpress LLC ADP-ribose-pNP disodium | 00-00-0 | 99.9% | 724.37 g/mol | C21H24N6Na2O16P2 | 5 MG
SDP
Catalog No. 5000222027
Encompass_Preferred
pNP-ADPr disodium is a colorimetric ADP-ribose p-nitrophenyl substrate used to measure poly(ADP-ribose) glycohydrolase (PARG) and ADP-ribosyl hydrolase 3 (ARH3) activity. It is supplied in small research pack sizes for enzymatic assays and enables continuous colorimetric monitoring of hydrolase activity.
  • Colorimetric substrate for poly(ADP-ribose) glycohydrolase (PARG) and ARH3 assays.
  • Enables the first continuous colorimetric assay format for PARG activity.
  • Suitable for kinetic and end-point enzymatic assays.
  • Supplied as the disodium salt for improved aqueous solubility.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • High analytical purity (~99.9%).
Catalog No. 50-002-22027 Supplier Medchemexpress LLC Supplier No. HY134354A5MG
pNP-ADPr disodium is a colorimetric ADP-ribose p-nitrophenyl substrate used to measure poly(ADP-ribose) glycohydrolase (PARG) and ADP-ribosyl hydrolase 3 (ARH3) activity. It is supplied in small research pack sizes for enzymatic assays and enables continuous colorimetric monitoring of hydrolase activity.
  • Colorimetric substrate for poly(ADP-ribose) glycohydrolase (PARG) and ARH3 assays.
  • Enables the first continuous colorimetric assay format for PARG activity.
  • Suitable for kinetic and end-point enzymatic assays.
  • Supplied as the disodium salt for improved aqueous solubility.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • High analytical purity (~99.9%).