Learn More
Chemglass Life Sciences Adapters, Inlet, 29/42 Inner Joint, Straight Swagelok, for 1/2" Compression Adapters
SDP
Catalog No. CG1012C02
Encompass_Preferred
Adapter, inlet, inner joint, straight. Joint Size: 29/42, Overall Length(mm): 95. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.
Catalog No. CG1012C02 Supplier Chemglass Life Sciences Supplier No. CG1012C02
Adapter, inlet, inner joint, straight. Joint Size: 29/42, Overall Length(mm): 95. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.