Learn More
Aviva Systems Biology Corp ADPGK Antibody - N-terminal region (ARP34632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADPGK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADPGK. Alternate names: 2610017G09Rik, ADP-GK, DKFZP434B195ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55491