Learn More
Aviva Systems Biology Corp ADPRH Antibody - C-terminal region (ARP51922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADPRH. It was validated on Western Blot. . Alternate names: ARH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-608