Learn More
Medchemexpress LLC HY-15904 500mg Medchemexpress, ADPS CAS:82611-88-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15904500MG
Encompass_Preferred
Medchemexpress, HY-15904 500mg ADPS CAS:82611-88-9 ADPS is a kind of new Trinder's reagents, which are high water-soluble aniline derivatives, widely used in diagnostic and biochemical tests. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1823