Learn More
IDEX Health & Science Upchurch ADPT 1/16IN BARBD-MLE 1/4IN-28
Catalog No. NC1919462
Encompass_Preferred
Idex Barbed-to-Threaded Adapters, Diameter Inner: 1.57 mm, ETFE, For Use With: Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports
Catalog No. NC1919462 Supplier IDEX Health & Science Upchurch Supplier No. P646
Idex Barbed-to-Threaded Adapters, Diameter Inner: 1.57 mm, ETFE, For Use With: Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports