Learn More
IDEX Health & Science Upchurch ADPT 3/16IN BARBD-MLE 1/4IN-28
Catalog No. NC1919466
Encompass_Preferred
Idex Barbed-to-Threaded Adapters, Diameter Inner: 4.76 mm, ETFE, For Use With: Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports
Catalog No. NC1919466 Supplier IDEX Health & Science Upchurch Supplier No. P648
Idex Barbed-to-Threaded Adapters, Diameter Inner: 4.76 mm, ETFE, For Use With: Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports