Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #50 Ace-Thred to 3/8in female NPT, 100micron polyethylene filter, 350 micron polypropylene filter, retaining ring, FETFE O-ring
SDP
Catalog No. NC1939718
Encompass_Preferred
Adapter,filter,PTFE,#50 Ace-Thred to 3/8in female NPT,100 micron polyethylene filter,350 micron polypropylene filter, retaining ring,Fetfe o-ring. ACE GLASS INC, mfg# 5838-80
Catalog No. NC1939718 Supplier Ace Glass, Inc. Supplier No. 583880
Adapter,filter,PTFE,#50 Ace-Thred to 3/8in female NPT,100 micron polyethylene filter,350 micron polypropylene filter, retaining ring,Fetfe o-ring. ACE GLASS INC, mfg# 5838-80