Learn More
Chemglass Life Sciences Adapters, Inlet, 45/50 Inner Joint, 90° Swagelok, for 1/2" Compression Adapters
SDP
Catalog No. CG1014C03
Encompass_Preferred
Adapter, Inlet, Joint Size: 45/50, Dimensions Height x Width (MM): 95 x 70. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.
Catalog No. CG1014C03 Supplier Chemglass Life Sciences Supplier No. CG1014C03
Adapter, Inlet, Joint Size: 45/50, Dimensions Height x Width (MM): 95 x 70. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.