Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #25 Ace-Thred to 1/4in female NPT, 100micron polyethylene filter, 350 micron polypropylene filter, retaining ring, FETFE O-ring
SDP
Catalog No. NC1905947
Encompass_Preferred
Adapter Ptfe #25-1/4In Npt. Ace Glass mfg# 5838-78
Catalog No. NC1905947 Supplier Ace Glass, Inc. Supplier No. 583878
Adapter Ptfe #25-1/4In Npt. Ace Glass mfg# 5838-78