Learn More
Chemglass Life Sciences Chemglass Adapter, 1-1/2 in Beaded Pipe to 3/4 in Compression Fitting, Glass-Filled PTFE, for Process Reactor Lower Outlet
SDP
Catalog No. NC2016070
Encompass_Preferred
Glass-filled PTFE adapter that converts a 1-1/2 in beaded reactor pipe connection to a 3/4 in compression fitting. Designed for use as a lower outlet on process and filter reactors, it provides a chemically resistant coupling for tubing and compression fittings.
  • Material: Glass-filled PTFE.
  • Beaded pipe size: 1-1/2 in.
  • Compression fitting size: 3/4 in.
  • Intended use: Adapter for process reactor lower outlets and filter reactor connections.
  • Compatibility: Requires a matching beaded pipe coupling.
  • Country of origin: Made in the United States.
Catalog No. NC2016070 Supplier Chemglass Life Sciences Supplier No. CG196455
Glass-filled PTFE adapter that converts a 1-1/2 in beaded reactor pipe connection to a 3/4 in compression fitting. Designed for use as a lower outlet on process and filter reactors, it provides a chemically resistant coupling for tubing and compression fittings.
  • Material: Glass-filled PTFE.
  • Beaded pipe size: 1-1/2 in.
  • Compression fitting size: 3/4 in.
  • Intended use: Adapter for process reactor lower outlets and filter reactor connections.
  • Compatibility: Requires a matching beaded pipe coupling.
  • Country of origin: Made in the United States.