Learn More
Chemglass Life Sciences Adapter, 60 mm Flat Flange Lid to 45/50 Outer Joint, Angled 20°
SDP
Catalog No. NC3002256
Encompass_Preferred
Glass adapter that connects a 60 mm flat flange lid to a 45/50 outer ground joint in an angled configuration, providing a sealed transition for vapors and accessory connections.
  • Converts 60 mm flat flange lid to a 45/50 outer joint
  • Angled 20° profile for directional vapor routing
  • Suitable for distillation and process reactor lids
  • Constructed from laboratory glassware for chemical compatibility
  • Provides sealed transition for vapor and accessory connections
Catalog No. NC3002256 Supplier Chemglass Life Sciences Supplier No. CG1965D205
Glass adapter that connects a 60 mm flat flange lid to a 45/50 outer ground joint in an angled configuration, providing a sealed transition for vapors and accessory connections.
  • Converts 60 mm flat flange lid to a 45/50 outer joint
  • Angled 20° profile for directional vapor routing
  • Suitable for distillation and process reactor lids
  • Constructed from laboratory glassware for chemical compatibility
  • Provides sealed transition for vapor and accessory connections