Learn More
Beckman Coulter Beckman Coulter 500 mL Conical Bottle Adapter for Allegra benchtop centrifuges (C63171)
Catalog No. NC2829984
Encompass_Preferred
Beckman Coulter's 500 mL conical bottle adapter (C63171) secures a single 500 mL conical bottle in a compatible centrifuge bucket or rotor, providing stable support during centrifugation and reducing bottle movement. It adapts rotor wells to accept conical bottles for processing culture bottles and similar containers.
  • Capacity: 500 mL
  • Holds one conical bottle per adapter
  • Designed for use with benchtop centrifuge buckets and compatible rotors
  • Secures bottles upright and centered during runs
  • Sold individually (1 bottle per adapter) according to manufacturer listing
Catalog No. NC2829984 Supplier Beckman Coulter Supplier No. C63171
Beckman Coulter's 500 mL conical bottle adapter (C63171) secures a single 500 mL conical bottle in a compatible centrifuge bucket or rotor, providing stable support during centrifugation and reducing bottle movement. It adapts rotor wells to accept conical bottles for processing culture bottles and similar containers.
  • Capacity: 500 mL
  • Holds one conical bottle per adapter
  • Designed for use with benchtop centrifuge buckets and compatible rotors
  • Secures bottles upright and centered during runs
  • Sold individually (1 bottle per adapter) according to manufacturer listing