Learn More
Chemglass Life Sciences Adapter, Distilling Trap, Kjeldahl, 14/20 Joint Size
SDP
Catalog No. NC1420896
Encompass_Preferred
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top standard taper outer and lower inner joints, both being the same size.
Catalog No. NC1420896 Supplier Chemglass Life Sciences Supplier No. CG103214
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam. With top standard taper outer and lower inner joints, both being the same size.