Learn More
Chemglass Life Sciences Adapter, Distillation, 24/40 Joint Size, Side Inner Joint At 75°
SDP
Catalog No. CG102201
Encompass_Preferred
Adapter, Distillation, 24/40 Joint Size, Side Inner Joint At 75¿, with standard taper outer joint at the top, and inner joints on the bottom and sidearm, all being the same size. Inner joints are at a 75¿ angle to one another.
Catalog No. CG102201 Supplier Chemglass Life Sciences Supplier No. CG102201
Adapter, Distillation, 24/40 Joint Size, Side Inner Joint At 75¿, with standard taper outer joint at the top, and inner joints on the bottom and sidearm, all being the same size. Inner joints are at a 75¿ angle to one another.