Learn More
Chemglass Life Sciences Adapter, Flow Control, 90°, 2mm Stpk, 24/40 Inner Joint
SDP
Supplier:  Chemglass Life Sciences CG103001
Encompass_Preferred
Adapter, Flow Control, 90¿, 2mm Stpk, 24/40 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm. Hose connection has an O.D. of 8mm at the middle serration.
Catalog No. CG103001