Learn More
Chemglass Life Sciences Adapter, Flow Control, 90°, 2mm Stpk, 24/40 Inner Joint
SDP
Catalog No. CG1030M01
Encompass_Preferred
Adapter, Flow Control, 90¿, 2mm Stpk, 24/40 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm, and has an internal needle valve which permits for fine control of gases or liquids added to reactions. Hose connection has an O.D. of 8mm at the middle serration.
Catalog No. CG1030M01 Supplier Chemglass Life Sciences Supplier No. CG1030M01
Adapter, Flow Control, 90¿, 2mm Stpk, 24/40 Inner Joint. Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2mm, and has an internal needle valve which permits for fine control of gases or liquids added to reactions. Hose connection has an O.D. of 8mm at the middle serration.