Learn More
Chemglass Life Sciences Adapter, Inlet, 14/20 Inner Joint, 55mm Height
SDP
Catalog No. CG101214
Encompass_Preferred
Adapter, Inlet, 14/20 Inner Joint, 55mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101214 Supplier Chemglass Life Sciences Supplier No. CG101214
Adapter, Inlet, 14/20 Inner Joint, 55mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.