Learn More
Chemglass Life Sciences Adapter, Inlet, 14/20 Outer Joint, 65mm Height
SDP
Catalog No. CG101314
Encompass_Preferred
Adapter, Inlet, 14/20 Outer Joint, 65mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101314 Supplier Chemglass Life Sciences Supplier No. CG101314
Adapter, Inlet, 14/20 Outer Joint, 65mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.