Learn More
Chemglass Life Sciences Adapter, Inlet, 24/40 Outer Joint, 95mm Height
SDP
Catalog No. CG101301
Encompass_Preferred
Adapter, Inlet, 24/40 Outer Joint, 95mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101301 Supplier Chemglass Life Sciences Supplier No. CG101301
Adapter, Inlet, 24/40 Outer Joint, 95mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.