Learn More
Chemglass Life Sciences Adapter, Thermometer, Inlet, 24/40 Inner Joint, 80mm Height, #7 Chem-Thread
SDP
Catalog No. CG104201
Encompass_Preferred
Adapter, Thermometer, Inlet, 24/40 Inner Joint, 80mm Height, #7 Chem-Thread. Adapter has a #7 or #15 Chem-Thread at the top for vacuum tight seal of plain stem thermo-meters or any other tube having an O.D. between 4 and 7mm. Supplied complete with a Celcon compression cap and Viton o-ring.
Catalog No. CG104201 Supplier Chemglass Life Sciences Supplier No. CG104201
Adapter, Thermometer, Inlet, 24/40 Inner Joint, 80mm Height, #7 Chem-Thread. Adapter has a #7 or #15 Chem-Thread at the top for vacuum tight seal of plain stem thermo-meters or any other tube having an O.D. between 4 and 7mm. Supplied complete with a Celcon compression cap and Viton o-ring.