Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Style "A", 105°, 24/40 Joint Size
SDP
Catalog No. CG105001
Encompass_Preferred
Adapter, Vacuum Take-Off, Style "A", 105¿, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. CG105001 Supplier Chemglass Life Sciences Supplier No. CG105001
Adapter, Vacuum Take-Off, Style "A", 105¿, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.