Learn More
FUJIFILM Biosciences ADRA Kit
Catalog No. 5000450397
Encompass_Preferred
Amino acid Derivative Reactivity Assay ADRA is an alternative method of skin sensitization test to animal testing ADRA significantly reduces the number of chemical substances used as a reaction reagent by using a novel amino acid derivative having a naphthalene ring capable of high sensitivity detection by ultraviolet rays Also skin sensitization tests are possible even at about 1/100 by conventional methods It is possible to evaluate skin sensitization with high accuracy without causing chemical substances to precipitate in the reaction solution It evaluates whether a chemical substance provokes an allergic reaction in the skin without using animal models Reagent kit for performing ADRA as described in OECD TG 442C
Catalog No. 50-004-50397 Supplier FUJIFILM Biosciences Supplier No. 29680901
Amino acid Derivative Reactivity Assay ADRA is an alternative method of skin sensitization test to animal testing ADRA significantly reduces the number of chemical substances used as a reaction reagent by using a novel amino acid derivative having a naphthalene ring capable of high sensitivity detection by ultraviolet rays Also skin sensitization tests are possible even at about 1/100 by conventional methods It is possible to evaluate skin sensitization with high accuracy without causing chemical substances to precipitate in the reaction solution It evaluates whether a chemical substance provokes an allergic reaction in the skin without using animal models Reagent kit for performing ADRA as described in OECD TG 442C