Learn More
Eurofins Discoverx Ready-to-Assay α1A Adrenergic Receptor Frozen Cells, 2 Vials of Cells. (HTS087RTA)
Catalog No. 502005839
Encompass
The Ready-to-Assay α1A Adrenergic Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1A alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of ADRA1A, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5839 Supplier Eurofins Discoverx Supplier No. HTS087RTA
The Ready-to-Assay α1A Adrenergic Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1A alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of ADRA1A, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.