Learn More
Aviva Systems Biology Corp ADRA1B Antibody - C-terminal region (ARP59960_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59960_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRA1B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ADRA1B. Alternate names: ADRA1, ALPHA1BARApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-898