Learn More
Aviva Systems Biology Corp ADRA1D Antibody - C-terminal region (ARP59958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRA1D. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ADRA1D. Alternate names: ADRA1, ADRA1A, ADRA1R, ALPHA1, DAR, dJ779E11.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-897