Learn More
Eurofins Discoverx ChemiScreen α1D Adrenergic Receptor with N-terminal truncation Membrane Preparation, 1 vial. (HTS216M)
Catalog No. 502006066
Encompass
The ChemiScreen α1D Adrenergic Receptor with N-terminal truncation Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of ADRA1D (82-572) surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-6066 Supplier Eurofins Discoverx Supplier No. HTS216M
The ChemiScreen α1D Adrenergic Receptor with N-terminal truncation Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of ADRA1D (82-572) surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.