Learn More
Eurofins Discoverx ChemiScreen α2A Adrenergic Receptor Stable Cell Line, 2 vials. (HTS096C)
Supplier:  Eurofins Discoverx HTS096C
Encompass
The ChemiScreen α2A Adrenergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA2A alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of ADRA2A, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5868
Add to Cart
Request a Quote