Learn More
Eurofins Discoverx ChemiBrite α2B Adrenergic Receptor Stable Cell Line, 2 vials. (HTS157L)
Catalog No. 502005979
Encompass
The ChemiBrite α2B Adrenergic Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2B GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA2B alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of ADRA2B, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5979 Supplier Eurofins Discoverx Supplier No. HTS157L
Add to Cart
Add to Cart
Request a Quote
The ChemiBrite α2B Adrenergic Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2B GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA2B alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of ADRA2B, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.