Learn More
Aviva Systems Biology Corp ADRB1 Antibody - middle region (ARP36551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADRB1. Alternate names: ADRB1R, B1AR, BETA1AR, RHRApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56185