Learn More
Aviva Systems Biology Corp ADRB2 Antibody - middle region (AVARP00006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADRB2. Alternate names: ADRB2R, ADRBR, B2AR, BAR, BETA2ARApplicationWB. Species reactivity: Human
Catalog No. 50-014-62444