Learn More
Enzo Life Sciences Adrenergic receptor beta2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905742100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the N-terminus of mouse adrenergic receptor β2. UniProt ID: P18762. Species reactivity: Human, Mouse, Rat. Creossreactivity: Predicted species reactivity based on epitope sequence homology: chimpanzee, orangutan, tamarin monkey, gorilla, rhesus macaque, horse, hamster, guinea pig, wild boar, cat. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-200-7030