Learn More
Eurofins Discoverx PathHunter ADRB2 Total Internalization HEK 293 Cell Line, 2 vials. (93-0800C1)
Catalog No. 502001323
Encompass
The PathHunter ADRB2 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of ADRB2 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged ADRB2 and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the ADRB2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1323 Supplier Eurofins Discoverx Supplier No. 930800C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter ADRB2 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of ADRB2 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged ADRB2 and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the ADRB2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.