Learn More
Eurofins Discoverx Ready-to-Assay β3 Adrenergic Receptor Frozen Cells, 2 Vials of Cells. (HTS159RTA)
Catalog No. 502005987
Encompass
The Ready-to-Assay β3 Adrenergic Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRB3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRB3 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of ADRB3, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5987 Supplier Eurofins Discoverx Supplier No. HTS159RTA
The Ready-to-Assay β3 Adrenergic Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRB3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRB3 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of ADRB3, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.