Learn More
Aviva Systems Biology Corp GRK2 Antibody - N-terminal region (ARP51965_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51965_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRBK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADRBK1. Alternate names: BARK1, BETA-ARK1, FLJ16718, GRK2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-63008