Learn More
Abcam Adrenaline ELISA Kit, 96TEST
Supplier:  Abcam AB28778896TEST
Encompass_Preferred
Adrenaline ELISA Kit ab287788 is used to quantify Adrenaline with a sensitivity of 2.1 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5885