Learn More
Selleck Chemical LLC Adrenalone HCl S3185-50mg
SDP
Supplier:  Selleck Chemical LLC S318550MG
Encompass_Preferred
Adrenalone is an adrenergic agonist used as a topical vasoconstrictor and hemostatic mainly acts on alpha-1 adrenergic receptors
Catalog No. 50-310-9859