Learn More
Apexbio Technology LLC Adrenalone HCl 62-13-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088705
Encompass_Preferred
Adrenalone hydrochloride (CAS 62-13-5) is a synthetic adrenergic agonist primarily targeting alpha-1 adrenergic receptors Through activation of these receptors Adrenalone induces vasoconstriction leading to reduced local blood flow This pharmacological property underpins its utility as a topical hemostatic agent in research settings where precise modulation of vascular tone is required Adrenalone is frequently employed to investigate adrenergic receptor signaling pathways and the physiological mechanisms underlying vascular constriction
Catalog No. 50-000-88705 Supplier Apexbio Technology LLC Supplier No. B13315.1
Adrenalone hydrochloride (CAS 62-13-5) is a synthetic adrenergic agonist primarily targeting alpha-1 adrenergic receptors Through activation of these receptors Adrenalone induces vasoconstriction leading to reduced local blood flow This pharmacological property underpins its utility as a topical hemostatic agent in research settings where precise modulation of vascular tone is required Adrenalone is frequently employed to investigate adrenergic receptor signaling pathways and the physiological mechanisms underlying vascular constriction