Learn More
Medchemexpress LLC Adrenalone hydrochloride | 62-13-5 | 99.88% | 500 MG
SDP
Catalog No. 5000411567
Encompass_Preferred
Adrenalone hydrochloride is an adrenergic agonist employed as a topical vasoconstrictor and hemostatic. It acts as an inhibitor of dopamine β oxidase and exhibits chemical similarity to known norepinephrine transporter (NET) ligands, with an IC50 of 36.9 μM. It is a keton form of the natural substrate epinephrine and also inhibits substrate transport by NET. It is used for research purposes only.
  • Acts as an adrenergic agonist
  • Functions as a topical vasoconstrictor
  • Exhibits hemostatic properties
  • Inhibits dopamine β oxidase
  • Chemically similar to norepinephrine transporter (NET) ligands
  • Available as a solid, light yellow to yellow in appearance
Catalog No. 50-004-11567 Supplier Medchemexpress LLC Supplier No. HYB1308500MG
Adrenalone hydrochloride is an adrenergic agonist employed as a topical vasoconstrictor and hemostatic. It acts as an inhibitor of dopamine β oxidase and exhibits chemical similarity to known norepinephrine transporter (NET) ligands, with an IC50 of 36.9 μM. It is a keton form of the natural substrate epinephrine and also inhibits substrate transport by NET. It is used for research purposes only.
  • Acts as an adrenergic agonist
  • Functions as a topical vasoconstrictor
  • Exhibits hemostatic properties
  • Inhibits dopamine β oxidase
  • Chemically similar to norepinephrine transporter (NET) ligands
  • Available as a solid, light yellow to yellow in appearance