Learn More
Cusabio Technology LLC adrencocorticotropic hormone
Catalog No. 5000049422
Encompass_Preferred
This kit detects ACTH (adrenocorticotropic hormone) in sheep serum and plasma samples using a competitive ELISA method Designed for quantitative analysis it offers a detection range of 0 097 pg/mL to 400 pg/mL with a sensitivity of 0 097 pg/mL enabling precise measurement of low-concentration samples The assay requires 50-100 L of sample volume and can be completed within 1-5 hours The optical density is measured at 450 nm to determine ACTH levels providing reliable results for research applications requiring adrenal function assessment in ovine species The kit s competitive binding format ensures specific detection while maintaining efficiency across biological matrices
Catalog No. 50-000-49422 Supplier Cusabio Technology LLC Supplier No. CSBEQ027618SH
This kit detects ACTH (adrenocorticotropic hormone) in sheep serum and plasma samples using a competitive ELISA method Designed for quantitative analysis it offers a detection range of 0 097 pg/mL to 400 pg/mL with a sensitivity of 0 097 pg/mL enabling precise measurement of low-concentration samples The assay requires 50-100 L of sample volume and can be completed within 1-5 hours The optical density is measured at 450 nm to determine ACTH levels providing reliable results for research applications requiring adrenal function assessment in ovine species The kit s competitive binding format ensures specific detection while maintaining efficiency across biological matrices