Learn More
Enzo Life Sciences SCREEN-WELL Adrenergic ligand library (500 µl/well)
SDP
Catalog No. 502010189
Encompass_Preferred
The adrenergic ligand library contains 83 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The adrenergic ligand library is 1 plate of the larger neurotransmitter library (BML-2810) which contains over 650 CNS receptor ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0189 Supplier Enzo Life Sciences Supplier No. BML28110500
The adrenergic ligand library contains 83 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The adrenergic ligand library is 1 plate of the larger neurotransmitter library (BML-2810) which contains over 650 CNS receptor ligands. Long Term Storage: -80°C. Applications: HTS .