Learn More
Cayman Chemical AdrenIc AcId 10mg
SDP
Catalog No. 502894859
Encompass_Preferred
A naturally occurring PUFA formed through a 2-carbon chain elongation of arachidonic acid; present in the adrenal glands, brain, testis, and kidney
Catalog No. 50-289-4859 Supplier Cayman Chemical Supplier No. 9030010 MG
A naturally occurring PUFA formed through a 2-carbon chain elongation of arachidonic acid; present in the adrenal glands, brain, testis, and kidney